Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000639.2 | NP_000630.1 | 1, 2 | 536 | ||
GenBank mRNA | AK314874.1 | - | 1, 2 | 500 | ||
AY225406.1 | - | 1, 2 | 342 | |||
AY858799.1 | - | 1, 2 | 342 | |||
BC017502.1 | - | 1, 2 | 429 | 4849770 | ||
D38122.1 | - | 1, 2 | 530 | |||
DA916336.1 | - | 1, 2 | 533 | |||
KJ948655.1 | - | 1, 2 | 499 | |||
U08137.1 | - | 1, 2 | 427 | |||
U11821.1 | - | 1, 2 | 406 | |||
X89102.1 | - | 1, 2 | 499 |