Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pole3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007652.2 | NP_001007653.1 | 3 | 196 | ||
GenBank mRNA | BC081988.1 | - | 3 | 196 | 7129121 | |
BC083800.1 | - | 3 | 196 | 7108773 |
Gene Symbol: LOC100362333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729829.4 | XP_002729875.1 | 1 | 257 |