Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206675.1 | NP_001193604.1 | 1 | 33 | ||
GenBank mRNA | FQ227858.1 | - | 1 | 69 |