Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp353 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_233185.7 | XP_233185.6 | 1 | 1802 |
Gene Symbol: LOC103694640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773747.1 | XP_008771969.1 | 1 | 73 |