Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134519.1 | NP_001127991.1 | 533 | |||
XM_006238584.2 | XP_006238646.1 | 970 | ||||
XM_006238585.2 | XP_006238647.1 | 560 | ||||
GenBank mRNA | BC168701.1 | - | 549 | 7454685 |
Gene Symbol: LOC100910316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749994.3 | XP_003750042.1 | 6, 7 | 1635 | ||
XM_006238582.2 | XP_006238644.1 | 6, 7 | 970 | |||
XM_008763953.1 | XP_008762175.1 | 6, 7 | 1276 |