Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320663.1 | NP_001307592.1 | 6 | 2003 | ||
NM_001320664.1 | NP_001307593.1 | 6 | 2000 | |||
NM_001649.3 | NP_001640.1 | 10 | 4768 | |||
XM_005274500.4 | XP_005274557.2 | 10 | 4815 | |||
XM_011545506.1 | XP_011543808.1 | 6 | 1880 | |||
XM_017029492.1 | XP_016884981.1 | 6 | 1877 | |||
XM_017029493.1 | XP_016884982.1 | 6 | 2373 | |||
XM_017029494.1 | XP_016884983.1 | 6 | 2370 | |||
GenBank mRNA | AK299907.1 | - | 6 | 2003 | ||
BC140866.1 | - | 10 | 4729 | 9021734 | ||
CR749271.1 | - | 6 | 1939 | |||
X83543.1 | - | 10 | 4768 |