Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256171.1 | NP_001243100.1 | 5 | 1377 | ||
NM_001256173.1 | NP_001243102.1 | 3 | 1160 | |||
NM_003429.4 | NP_003420.2 | 4 | 1287 | |||
XM_011528263.2 | XP_011526565.1 | 6 | 1448 | |||
XM_011528264.1 | XP_011526566.1 | 2 | 1608 | |||
XM_017027234.1 | XP_016882723.1 | 2 | 1087 | |||
GenBank mRNA | AK308258.1 | - | 4 | 1117 | ||
BC047646.1 | - | 2 | 1590 | 4828290 | ||
D70831.1 | - | 1 | 686 | |||
U35376.1 | - | 4 | 1257 |