Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000412.1 | NP_001000412.1 | 1 | 835 |
Gene Symbol: Olfr1330-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238754.1 | XP_006238816.1 | 2 | 910 |
Gene Symbol: Olr860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000822.1 | NP_001000822.1 | 1 | 835 |
Gene Symbol: Olr862 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001071.1 | NP_001001071.1 | 1 | 829 |
Gene Symbol: Olr865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000410.1 | NP_001000410.1 | 1 | 835 | ||
XM_008763996.1 | XP_008762218.1 | 1 | 835 |
Gene Symbol: Olr866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000411.1 | NP_001000411.1 | 1 | 835 |
Gene Symbol: Olr868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000821.1 | NP_001000821.1 | 1 | 835 |
Gene Symbol: Olr869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000413.1 | NP_001000413.1 | 1 | 835 |