Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000413.1 | NP_001000413.1 | 1 | 529 |
Gene Symbol: Olfr1330-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238754.1 | XP_006238816.1 | 2 | 604 |
Gene Symbol: Olr867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000412.1 | NP_001000412.1 | 1 | 529 |