Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106683.1 | NP_001100153.1 | 5 | 682 | ||
XM_006238792.2 | XP_006238854.1 | 5 | 662 | |||
XM_008764001.1 | XP_008762223.1 | 5 | 659 | |||
GenBank mRNA | BC161915.1 | - | 5 | 669 | 8369688 |