Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106686.1 | NP_001100156.1 | 10 | 1298 | ||
XM_006238862.2 | XP_006238924.1 | 10 | 1570 |