Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201407.1 | NP_001188336.1 | 7 | 2477 | ||
NM_182531.3 | NP_872337.2 | 6 | 2393 | |||
NR_037705.1 | 6 | 2350 | ||||
XM_005256288.2 | XP_005256345.1 | 7 | 2463 | |||
XM_011522940.2 | XP_011521242.1 | 7 | 2430 | |||
XM_017023015.1 | XP_016878504.1 | 6 | 2385 | |||
XM_017023016.1 | XP_016878505.1 | 7 | 2557 | |||
XM_017023017.1 | XP_016878506.1 | 7 | 2456 | |||
GenBank mRNA | AK056437.1 | - | 6 | 2385 | ||
BC015872.1 | - | 1 | 725 | |||
BC125192.1 | - | 5 | 2098 | 40122711 |
Gene Symbol: ZNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008727.3 | NP_001008727.1 | 4 | 951 | ||
NM_001308269.1 | NP_001295198.1 | 5 | 1009 | |||
XM_017027239.1 | XP_016882728.1 | 5 | 1125 | |||
GenBank mRNA | AK297919.1 | - | 5 | 1008 | ||
AW949690.1 | - | 1 | 61 | |||
CN360955.1 | - | 1 | 366 |