Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106688.1 | NP_001100158.1 | 1, 2 | 175 | ||
GenBank mRNA | BC167002.1 | - | 1, 2 | 100 | 7443117 |