Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013935.1 | NP_001013957.1 | 6 | 673 | ||
XM_006239124.2 | XP_006239186.1 | 6 | 969 | |||
XM_006239125.2 | XP_006239187.1 | 6 | 747 | |||
GenBank mRNA | BC083692.1 | - | 6 | 673 | 7189448 |