Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005544.1 | NP_001005544.1 | 4 | 264 | ||
XM_008764346.1 | XP_008762568.1 | 4 | 335 | |||
XM_008764347.1 | XP_008762569.1 | 3 | 236 | |||
XM_008764348.1 | XP_008762570.1 | 4 | 335 | |||
XM_008764349.1 | XP_008762571.1 | 3 | 174 | |||
XM_008764350.1 | XP_008762572.1 | 3 | 210 | |||
GenBank mRNA | BC082091.1 | - | 4 | 264 | 7134071 |