Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034922.1 | NP_001030094.1 | 14 | 1509 | ||
GenBank mRNA | BC105762.1 | - | 14 | 1509 | 7929859 |