Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF133 SIRNA Assays ›
Gene Symbol:
ZNF133
Gene Name:
zinc finger protein 133
Gene Aliases:
ZNF150, pHZ-13, pHZ-66
Chromosome Location:
Chr.20: 18288228 - 18316996 on Build GRCh38
UniGene ID:
Hs.472221
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15166
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF133
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083330.2
NP_001076799.1
4
2120
NM_001282995.1
NP_001269924.1
6
2356
NM_001282996.1
NP_001269925.1
6
2230
NM_001282997.1
NP_001269926.1
5
2281
NM_001282998.1
NP_001269927.1
4
2110
NM_001282999.1
NP_001269928.1
4
2123
NM_001283000.1
NP_001269929.1
5
2299
NM_001283001.1
NP_001269930.1
5
2286
NM_001283002.1
NP_001269931.1
5
2188
NM_001283003.1
NP_001269932.1
5
2261
NM_001283004.1
NP_001269933.1
5
2248
NM_001283005.1
NP_001269934.1
2
1884
NM_001283006.1
NP_001269935.1
3
1962
NM_001283007.1
NP_001269936.1
3
1980
NM_001283008.1
NP_001269937.1
4
2150
NM_003434.5
NP_003425.2
7
2532
XM_005260819.1
XP_005260876.1
4
1932
XM_005260820.4
XP_005260877.1
6
2189
XM_011529336.1
XP_011527638.1
7
2268
XM_011529337.1
XP_011527639.1
6
2140
XM_011529338.2
XP_011527640.1
6
2134
XM_011529339.1
XP_011527641.1
7
2268
XM_011529340.1
XP_011527642.1
2
1743
XM_017028041.1
XP_016883530.1
5
10033
XM_017028042.1
XP_016883531.1
5
3307
XM_017028043.1
XP_016883532.1
5
10030
XM_017028044.1
XP_016883533.1
7
10380
XM_017028045.1
XP_016883534.1
5
10448
XM_017028046.1
XP_016883535.1
5
10046
XM_017028047.1
XP_016883536.1
5
3400
XM_017028048.1
XP_016883537.1
4
3229
XM_017028049.1
XP_016883538.1
7
10390
XM_017028050.1
XP_016883539.1
7
2288
XM_017028051.1
XP_016883540.1
8
2464
XM_017028052.1
XP_016883541.1
6
2128
XM_017028053.1
XP_016883542.1
4
2162
XM_017028054.1
XP_016883543.1
5
2042
XM_017028055.1
XP_016883544.1
8
2458
XM_017028056.1
XP_016883545.1
10
2677
XM_017028057.1
XP_016883546.1
7
2245
XM_017028058.1
XP_016883547.1
8
2462
GenBank mRNA
AK123005.1
-
2
3698
AK291389.1
-
5
2038
AK294263.1
-
3
1711
AK295273.1
-
5
1960
AK295513.1
-
5
2024
AK313525.1
-
4
1859
AK316386.1
-
2
1638
BC001887.1
-
7
2338
3535533
BT007310.1
-
3
1779
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline