Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013095.1 | NP_001013113.1 | 10 | 933 | ||
GenBank mRNA | BC086383.1 | - | 10 | 933 | 7301572 | |
FQ213005.1 | - | 10 | 974 | |||
FQ225883.1 | - | 10 | 966 | |||
FQ226276.1 | - | 10 | 965 | |||
FQ229703.1 | - | 10 | 964 | |||
FQ230888.1 | - | 10 | 959 | |||
FQ232595.1 | - | 10 | 963 | |||
FQ232661.1 | - | 10 | 963 | |||
FQ234018.1 | - | 10 | 967 | |||
FQ234727.1 | - | 8 | 815 | |||
FQ234921.1 | - | 10 | 964 | |||
FQ234929.1 | - | 10 | 966 | |||
FQ235264.1 | - | 7 | 747 |