Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raver1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013939.1 | NP_001013961.1 | 10 | 1851 | ||
GenBank mRNA | BC083865.1 | - | 10 | 1851 | 7127766 |