Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013940.1 | NP_001013962.1 | 5 | 383 | ||
XM_006239695.2 | XP_006239757.1 | 5 | 438 | |||
GenBank mRNA | BC079201.1 | - | 5 | 383 | 7131728 |