Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106706.1 | NP_001100176.1 | 12 | 1562 | ||
XM_006239754.2 | XP_006239816.1 | 12 | 1580 |