Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199384.2 | NP_955416.1 | 5, 6 | 601 | ||
GenBank mRNA | BC063179.1 | - | 5, 6 | 605 | 6890564 | |
FQ212732.1 | - | 5, 6 | 1003 | |||
FQ213363.1 | - | 5, 6 | 635 | |||
FQ229677.1 | - | 560 | ||||
FQ232171.1 | - | 5, 6 | 659 | |||
FQ232930.1 | - | 5, 6 | 619 | |||
FQ235113.1 | - | 5, 6 | 506 |