Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282656.1 | NP_001269585.1 | 13, 14 | 1645 | ||
NM_001282657.1 | NP_001269586.1 | 12, 13 | 1555 | |||
NM_003442.5 | NP_003433.3 | 13, 14 | 1648 | |||
XM_005253121.1 | XP_005253178.1 | 13, 14 | 1596 | |||
XM_005253122.1 | XP_005253179.1 | 13, 14 | 1621 | |||
XM_005253127.1 | XP_005253184.1 | 14, 15 | 1674 | |||
XM_011520349.2 | XP_011518651.1 | 13, 14 | 2367 | |||
XM_017018252.1 | XP_016873741.1 | 12, 13 | 1532 | |||
XM_017018253.1 | XP_016873742.1 | 12, 13 | 1529 | |||
XM_017018254.1 | XP_016873743.1 | 13, 14 | 1633 | |||
XM_017018255.1 | XP_016873744.1 | 13, 14 | 1723 | |||
XM_017018256.1 | XP_016873745.1 | 12, 13 | 1535 | |||
GenBank mRNA | AF071771.1 | - | 10, 11 | 1268 | ||
AK291133.1 | - | 13, 14 | 1601 | |||
AK297214.1 | - | 12, 13 | 1500 | |||
AK313330.1 | - | 13, 14 | 1634 | |||
BC020219.1 | - | 13, 14 | 1647 | 4902668 |