Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106740.1 | NP_001100210.1 | 3 | 831 | ||
XM_006240222.2 | XP_006240284.1 | 3 | 881 | |||
XM_006240223.2 | XP_006240285.1 | 3 | 879 | |||
XM_006240224.2 | XP_006240286.1 | 2 | 619 | |||
XM_006240225.2 | XP_006240287.1 | 2 | 528 |