Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106742.1 | NP_001100212.1 | 6, 7 | 650 | ||
GenBank mRNA | BC158786.1 | - | 6, 7 | 580 | 7303059 |