Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acyp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106746.2 | NP_001100216.1 | 3 | 399 | ||
NM_001199167.1 | NP_001186096.1 | 3 | 331 | |||
NM_001199168.1 | NP_001186097.1 | 3 | 295 | |||
XM_006240308.2 | XP_006240370.1 | 3 | 450 | |||
GenBank mRNA | FM034312.1 | - | 3 | 295 |