Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Batf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106748.1 | NP_001100218.1 | 3 | 435 | ||
XM_008764758.1 | XP_008762980.1 | 4 | 537 |