Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287603.1 | NP_001274532.1 | 3 | 334 | ||
NM_001287604.1 | NP_001274533.1 | 3 | 334 | |||
NM_001324138.1 | NP_001311067.1 | 3 | 334 | |||
NM_003443.2 | NP_003434.2 | 3 | 334 | |||
XM_005245986.1 | XP_005246043.1 | 3 | 334 | |||
XM_005245987.1 | XP_005246044.1 | 3 | 279 | |||
XM_011542086.1 | XP_011540388.1 | 3 | 279 | |||
XM_011542088.1 | XP_011540390.1 | 3 | 279 | |||
XM_017002242.1 | XP_016857731.1 | 1 | 97 | |||
GenBank mRNA | AK223618.1 | - | 3 | 311 | ||
AK226168.1 | - | 3 | 274 | |||
AK294796.1 | - | 3 | 324 | |||
AK299820.1 | - | 3 | 291 | |||
AK303823.1 | - | 3 | 283 | |||
AK309081.1 | - | 2 | 196 | |||
AK311083.1 | - | 3 | 281 | |||
AK315201.1 | - | 3 | 281 | |||
BC126163.1 | - | 2 | 197 | 8991879 | ||
BC143965.1 | - | 2 | 197 | 9052483 | ||
DB064089.1 | - | 3 | 334 | |||
Y09723.1 | - | 3 | 221 |