Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106754.1 | NP_001100224.1 | 2 | 231 | ||
XM_008764764.1 | XP_008762986.1 | 2 | 211 | |||
XM_008764765.1 | XP_008762987.1 | 2 | 206 |