Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270982.1 | NP_001257911.1 | 2 | 419 | ||
XM_006240466.2 | XP_006240528.1 | 2 | 420 | |||
GenBank mRNA | FQ211337.1 | - | 1 | 102 | ||
X69834.1 | - | 1 | 298 |