Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC299282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182474.1 | NP_872280.1 | 10 | 1761 | ||
GenBank mRNA | AY298742.1 | - | 10 | 1761 | ||
AY325133.1 | - | 10 | 1761 |