Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_216781.8 | XP_216781.6 | 6 | 819 | ||
GenBank mRNA | BC089108.1 | - | 1 | 219 |
Gene Symbol: LOC100910833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764908.1 | XP_008763130.1 | 3 | 2222 |