Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764983.1 | XP_008763205.1 | 33, 34 | 3989 | ||
XM_008764984.1 | XP_008763206.1 | 33, 34 | 3989 | |||
XM_008764985.1 | XP_008763207.1 | 33, 34 | 3956 | |||
XM_008764986.1 | XP_008763208.1 | 33, 34 | 3956 | |||
XM_008764987.1 | XP_008763209.1 | 32, 33 | 3872 | |||
XM_008764988.1 | XP_008763210.1 | 33, 34 | 3760 | |||
GenBank mRNA | BC087164.1 | - | 24, 25 | 4134 | ||
BC104675.1 | - | 10, 11 | 1950 |