Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1075 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000421.1 | NP_001000421.1 | 1 | 72 |
Gene Symbol: Olr1076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000712.1 | NP_001000712.1 | 1 | 72 |