Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106768.1 | NP_001100238.1 | 2 | 241 | ||
XM_006240861.2 | XP_006240923.1 | 2 | 243 | |||
XM_006240862.2 | XP_006240924.1 | 2 | 240 | |||
XM_006240864.2 | XP_006240926.1 | 2 | 240 | |||
XM_006240865.2 | XP_006240927.1 | 2 | 236 | |||
XM_006240866.2 | XP_006240928.1 | 2 | 232 |