Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241035.2 | XP_006241097.1 | 5 | 2262 | ||
XM_008765148.1 | XP_008763370.1 | 9 | 2263 | |||
XM_008765149.1 | XP_008763371.1 | 9 | 2534 | |||
XM_216850.8 | XP_216850.6 | 8 | 2675 | |||
GenBank mRNA | FQ212316.1 | - | 8 | 1499 |