Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102381.1 | NP_001095851.1 | 4 | 513 | ||
GenBank mRNA | FQ214250.1 | - | 4 | 586 |