Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198751.1 | NP_942046.1 | 165 | |||
GenBank mRNA | BC060586.1 | - | 165 | 6918037 | ||
FQ216838.1 | - | 162 | ||||
FQ220633.1 | - | 164 | ||||
FQ228544.1 | - | 148 | ||||
FQ229203.1 | - | 158 | ||||
FQ232428.1 | - | 163 |
Gene Symbol: LOC100911110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750101.3 | XP_003750149.1 | 1, 2 | 165 |