Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130545.1 | NP_001124017.1 | 3 | 499 | ||
XM_006241726.2 | XP_006241788.1 | 3 | 687 |