Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134839.1 | NP_001128311.1 | 4 | 453 | ||
XM_006241781.2 | XP_006241843.1 | 4 | 477 | |||
XM_006241782.2 | XP_006241844.1 | 5 | 663 | |||
XM_006241783.2 | XP_006241845.1 | 4 | 402 | |||
GenBank mRNA | BC169114.1 | - | 4 | 435 |