Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317384.1 | NP_001304313.1 | 3 | 750 | ||
NM_001317387.1 | NP_001304316.1 | 3 | 750 | |||
NM_001650.5 | NP_001641.1 | 3 | 750 | |||
NM_004028.4 | NP_004019.1 | 2 | 877 | |||
XM_011525942.2 | XP_011524244.1 | 3 | 792 | |||
GenBank mRNA | AB128929.1 | - | 4 | 280 | ||
AK222684.1 | - | 3 | 662 | |||
AK295069.1 | - | 4 | 561 | |||
BC022286.1 | - | 3 | 615 | 4717755 | ||
D63412.1 | - | 3 | 615 | |||
HQ901095.1 | - | 3 | 724 | |||
KF055862.1 | - | 3 | 595 | |||
U34845.1 | - | 2 | 813 | |||
U63622.1 | - | 3 | 634 | |||
U63623.1 | - | 2 | 538 |