Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130493.1 | NP_001123965.1 | 3 | 1060 | ||
XM_008765565.1 | XP_008763787.1 | 3 | 1297 | |||
GenBank mRNA | BC158731.1 | - | 3 | 970 |