Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034135.1 | NP_001029307.1 | 117 | |||
GenBank mRNA | BC100258.1 | - | 117 | 7444286 |