Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004242.1 | NP_001004242.1 | 12 | 1320 | ||
GenBank mRNA | BC079089.1 | - | 12 | 1320 | 7123477 |