Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130500.1 | NP_001123972.1 | 6 | 1345 | ||
XM_006242160.2 | XP_006242222.1 | 7 | 1449 | |||
GenBank mRNA | FQ218328.1 | - | 7 | 1417 |