Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271173.1 | NP_001258102.1 | 25 | 3330 | ||
XM_006242184.2 | XP_006242246.1 | 25 | 3383 | |||
XM_006242185.2 | XP_006242247.1 | 23 | 2990 |