Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc48a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127456.1 | NP_001120928.1 | 2, 3 | 358 | ||
GenBank mRNA | BC105856.1 | - | 1, 2 | 271 | ||
BC158869.1 | - | 2, 3 | 358 | 7930286 |