Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765844.1 | XP_008764066.1 | 9 | 1418 | ||
XM_008765845.1 | XP_008764067.1 | 9 | 1264 | |||
XM_008765846.1 | XP_008764068.1 | 8 | 1382 | |||
XM_008765847.1 | XP_008764069.1 | 9 | 1418 | |||
XM_008765848.1 | XP_008764070.1 | 9 | 891 | |||
XM_008765849.1 | XP_008764071.1 | 8 | 1382 | |||
XM_008765850.1 | XP_008764072.1 | 9 | 891 | |||
GenBank mRNA | FQ230816.1 | - | 8 | 814 |
Gene Symbol: LOC103692995 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765881.1 | XP_008764103.1 | 3 | 347 |