Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011995.1 | NP_001011995.1 | 4 | 1502 | ||
GenBank mRNA | BC078829.1 | - | 4 | 1502 | 7112420 | |
FQ221879.1 | - | 3 | 1293 | |||
FQ225183.1 | - | 4 | 1523 | |||
FQ225366.1 | - | 4 | 1524 | |||
FQ226218.1 | - | 4 | 1523 | |||
FQ233955.1 | - | 2 | 1082 | |||
FQ235315.1 | - | 4 | 1526 |
Gene Symbol: LOC100909441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749354.3 | XP_003749402.1 | 4 | 1513 |