Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Troap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271243.1 | NP_001258172.1 | 12 | 1381 | ||
XM_006257348.2 | XP_006257410.1 | 11 | 1274 |